2-Methyl-1-(p-tolyl)-2-propanol 95%
Catalog # APOSOR930931-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:164.25 g/mol
Storage Temperature:Ambient
CAS Number:20834-59-7
:MFCD00021809
Formula:C₁₁H₁₆O
MDL Number:MFCD00029997