4-(Chloromethyl)-1-methylpiperidine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1182284-45-2
:
Formula:C₇H₁₅Cl₂N
MDL Number:MFCD12923013
:184.11 g/mol
Storage Temperature:Ambient
Frequently Bought Together