4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5451-55-8
:
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00042622
:184.28 g/mol
Storage Temperature:Ambient
Frequently Bought Together