
Tetrahydro-4H-pyran-4-one 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29943-42-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00006581
:100.12 g/mol
Boiling Pt:165…167 °C
Density:1.084 g/cm³ (25 °C)
Flash Pt:55 °C
Storage Temperature:Ambient
EINECS:249-967-2
UN:1224
ADR:3,III
:GHS02
Frequently Bought Together