
Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-4-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:189512-01-4
:
Formula:C₁₁H₁₆N₂O₄S
MDL Number:MFCD09908086
:272.33 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together