Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride 96%
  APOSOR925859-5G
 :  
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride 96%
  APOSOR925859-5G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

 :
 :193.67 g/mol
 :Ambient
 :61367-16-6
 :C₈H₁₅NO₂
 :MFCD08274537