Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6,7,8-Tetrahydro-2H-cinnolin-3-one
5,6,7,8-Tetrahydro-2H-cinnolin-3-one 95%
5,6,7,8-Tetrahydro-2H-cinnolin-3-one 95%
APOSOR925712-1G
:
Apollo Scientific
:
5,6,7,8-Tetrahydro-2H-cinnolin-3-one 95%
APOSOR925712-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
150.18 g/mol
:
Ambient
:
5468-36-0
:
C₈H₁₀N₂O
:
MFCD00828986