Order Entry
Czech Republic
ContactUsLinkComponent
(R)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
(R)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
Catalog # APOSOR925276-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
Catalog # APOSOR925276-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.24 g/mol
Storage Temperature:Ambient
CAS Number:100570-24-9
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD06761897