Methyl-4-(2-hydroxyethoxy)benzoate 95%
Catalog # APOSOR924639-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.20 g/mol
Storage Temperature:Ambient
CAS Number:3204-73-7
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00075872