Order Entry
Czech Republic
ContactUsLinkComponent
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) 98%
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) 98%
Catalog # APOSOR924567-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Methyl-1-cyclohexanemethanol (cis- and trans- mixture) 98%
Catalog # APOSOR924567-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :128.22 g/mol
Storage Temperature:Ambient
CAS Number:34885-03-5
Formula:C₈H₁₆O
MDL Number:MFCD00152628