Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98%
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98%
APOSOR920038-5G
:
Apollo Scientific
:
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol 98%
APOSOR920038-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
263.43 g/mol
:
GHS07
:
Ambient
:
114389-70-7
:
C₁₇H₂₉NO
:
MFCD00142695