Order Entry
Czech Republic
ContactUsLinkComponent
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine 98%
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine 98%
Catalog # APOSOR918240-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine 98%
Catalog # APOSOR918240-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.62 g/mol
Storage Temperature:Ambient
CAS Number:1060801-42-4
Formula:C₇H₉ClN₂
MDL Number:MFCD13188973