
(4-Chloro-1H-pyrazol-1-yl)acetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113336-23-5
:
Formula:C₅H₄ClN₃
MDL Number:MFCD04969697
:141.56 g/mol
Storage Temperature:Refrigerator
UN:3439
ADR:6.1,III
:GHS06
Frequently Bought Together