Order Entry
Czech Republic
ContactUsLinkComponent
4-Isopropyl-m-cresol 95%
Irritant
4-Isopropyl-m-cresol 95%
Catalog # APOSOR917093-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Isopropyl-m-cresol 95%
Catalog # APOSOR917093-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :150.22
Melting Pt:111…114 °C
EINECS:221-761-7
 :GHS07
Storage Temperature:Ambient
CAS Number:3228-02-2
Formula:C₁₀H₁₄O
MDL Number:MFCD00010704