2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture) 95%
Catalog # APOSOR916743-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.26 g/mol
Storage Temperature:Ambient
CAS Number:1052223-70-7
Formula:C₉H₁₉NO
MDL Number:MFCD07772981