Order Entry
Czech Republic
ContactUsLinkComponent
3-Methyl-2-hexanone 95%
3-Methyl-2-hexanone 95%
Catalog # APOSOR916391-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-2-hexanone 95%
Catalog # APOSOR916391-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :114.19 g/mol
Storage Temperature:Ambient
CAS Number:2550-21-2
Formula:C₇H₁₄O
MDL Number:MFCD00039948