Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-5-methylpyrazin-2-amine
3-Bromo-5-methylpyrazin-2-amine 95%
3-Bromo-5-methylpyrazin-2-amine 95%
APOSOR914309-1G
:
Apollo Scientific
:
3-Bromo-5-methylpyrazin-2-amine 95%
APOSOR914309-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
188.0300 g/mol
:
Ambient
:
74290-65-6
:
C5H6BrN3
:
MFCD09750037