(S)-(-)-2-Methyl-1,4-butanediol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:70423-38-0
:
Formula:C₅H₁₂O₂
MDL Number:MFCD00671566
Storage Temperature:Ambient
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Laboratory bottles, premium, with screw caps, DURAN®
Supplier: DWK Life Sciences