Benzyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:315718-05-9
:
Formula:C₁₃H₁₇NO₃
MDL Number:MFCD02179041
:235.29 g/mol
Storage Temperature:Ambient
Frequently Bought Together