Order Entry
Czech Republic
ContactUsLinkComponent
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 95%
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 95%
Catalog # APOSOR911717-100G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) 95%
Catalog # APOSOR911717-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :130.14 g/mol
Storage Temperature:Ambient
CAS Number:332-77-4
Formula:C₆H₁₀O₃
MDL Number:MFCD00003220