
Di-tert-butyl iminodicarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51779-32-9
:
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD00043309
:217.27 g/mol
Melting Pt:114…117 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together