Order Entry
Czech Republic
ContactUsLinkComponent
6-Methyl-3-nitroimidazo[1,2-a]pyridine 98%
6-Methyl-3-nitroimidazo[1,2-a]pyridine 98%
Catalog # APOSOR908963-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-3-nitroimidazo[1,2-a]pyridine 98%
Catalog # APOSOR908963-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.160 g/mol
Storage Temperature:Ambient
CAS Number:67625-28-9
Formula:C₈H₇N₃O₂
MDL Number:MFCD06496196