Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate 98%
Catalog # APOSOR908135-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate 98%
Catalog # APOSOR908135-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.19 g/mol
Storage Temperature:Ambient
CAS Number:214894-91-4
Formula:C₁₀H₁₀O₄
MDL Number:MFCD01546256