Tetrahydropyran-2-carboxylic acid 97%
Catalog # APOSOR907468-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:130.1418 g/mol
Storage Temperature:Ambient
CAS Number:51673-83-7
Formula:C₆H₁₀O₃
MDL Number:MFCD07779239