Order Entry
Czech Republic
ContactUsLinkComponent
N-Butyl-4-piperidinecarboxamide hydrochloride 95%
N-Butyl-4-piperidinecarboxamide hydrochloride 95%
Catalog # APOSOR907232-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Butyl-4-piperidinecarboxamide hydrochloride 95%
Catalog # APOSOR907232-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.74 g/mol
Storage Temperature:Ambient
CAS Number:73415-55-1
Formula:C₁₀H₂₁ClN₂O
MDL Number:MFCD11615740