Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-bromo-4-methylpyrimidine
2-Amino-5-bromo-4-methylpyrimidine 98%
2-Amino-5-bromo-4-methylpyrimidine 98%
APOSOR906853-1G
:
Apollo Scientific
:
2-Amino-5-bromo-4-methylpyrimidine 98%
APOSOR906853-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
188.03 g/mol
:
192…194 °C
:
GHS05
:
GHS07
:
Refrigerator
:
17321-93-6
:
C₅H₆BrN₃
:
MFCD00094469