
tert-Butyl-2-ethyl-4-oxopiperidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:324769-07-5
:
Formula:C₁₂H₂₁NO₃
MDL Number:MFCD07367729
:227.31 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together