7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24415-66-5
:
Formula:C₆H₅ClN₄
MDL Number:MFCD00227619
:168.5837 g/mol
Storage Temperature:Ambient
Frequently Bought Together