2,3,4,5-Tetrahydro-1H-benzo[c]azepine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17724-36-6
:
Formula:C₁₀H₁₄ClN
MDL Number:MFCD12198423
:183.6800 g/mol
Storage Temperature:Ambient
Frequently Bought Together