Order Entry
Czech Republic
ContactUsLinkComponent
6-Methoxy-m-toluidine 98%
6-Methoxy-m-toluidine 98%
Catalog # APOSOR905244-100G
Supplier:  Apollo Scientific
CAS Number:  
6-Methoxy-m-toluidine 98%
Catalog # APOSOR905244-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :137.18 g/mol
Storage Temperature:Ambient
CAS Number:120-71-8
Formula:C₈H₁₁NO
MDL Number:MFCD00007815