Order Entry
Czech Republic
ContactUsLinkComponent
2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride 95%
2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride 95%
Catalog # APOSOR905048-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride 95%
Catalog # APOSOR905048-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.64 g/mol
Storage Temperature:Ambient
CAS Number:173159-45-0
Formula:C₈H₁₀ClN₃
MDL Number:MFCD07367043