4-Chloro-6,7-dihydro-5H-pyrrolo[2,3-d]pyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16372-08-0
:
Formula:C₆H₆ClN₃
MDL Number:MFCD11518925
:155.5849 g/mol
Storage Temperature:Ambient
Frequently Bought Together