4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:126689-00-7
:
Formula:C₁₀H₁₉BO₂
MDL Number:MFCD09842762
:182.07 g/mol
Storage Temperature:Ambient
Frequently Bought Together