
m-Toluenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1899-93-0
:
Formula:C₇H₇ClO₂S
MDL Number:MFCD00051747
Boiling Pt:252…253 °C
Density:1.314 g/cm³ (25 °C)
Flash Pt:107.2 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:190.65 g/mol
:GHS05
Frequently Bought Together