Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

4-Ethoxybenzonitrile 98%

Dodavatel:   Apollo Scientific
4-Ethoxybenzonitrile 98%
Dodavatel:  Apollo Scientific
Certifikáty
4-Ethoxybenzonitrile 98%

5 Dostupných variant níže

Product Details & Documents
Synonyma:  p-Ethoxybenzonitrile
  • Číslo CAS:25117-74-2
  • Vzorec:C₉H₉NO
  • MDL číslo:MFCD00001819
  • Molecular Weight:147.18 g/mol
  • Teplota pro skladování:Ambient
Product Family Options

Informace o produktu

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.