
4-Isopropylbenzenesulphonyl chloride, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54997-90-9
:
Formula:C₉H₁₁ClO₂S
MDL Number:MFCD00041510
:218.7 g/mol
Boiling Pt:142 °C/5mmHg
Density:1.22 g/cm³ (25 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together