
Diethyl-2-chloro-3-oxosuccinate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:34034-87-2
:
Formula:C₈H₁₁ClO₅
MDL Number:MFCD04114359
:222.6229 g/mol
Boiling Pt:98 °C/0.5mmHg
Density:1.23 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together