
2',6'-Dihydroxyacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:699-83-2
:
Formula:C₈H₈O₃
MDL Number:MFCD00002270
:152.15 g/mol
Melting Pt:156…159 °C
EINECS:211-833-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together