
2-Methoxy-4-nitrobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21320-91-2
:
Formula:C₇H₆ClNO₅S
MDL Number:MFCD03094697
Melting Pt:90…99 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together