Order Entry
Czech Republic
ContactUsLinkComponent
2-Cyanobenzenesulphonyl chloride
Corrosive
2-Cyanobenzenesulphonyl chloride
Catalog # APOSOR8940-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Cyanobenzenesulphonyl chloride
Catalog # APOSOR8940-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:65…69 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :201.63 g/mol
 :GHS05
CAS Number:69360-26-5
Formula:C₇H₄ClNO₂S
MDL Number:MFCD00052341