Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-1-cyclohexene-1-carboxylate 98%
Methyl-1-cyclohexene-1-carboxylate 98%
Catalog # APOSOR8858-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-1-cyclohexene-1-carboxylate 98%
Catalog # APOSOR8858-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :140.182 g/mol
Storage Temperature:Ambient
CAS Number:18448-47-0
Formula:C₈H₁₂O₂
MDL Number:MFCD00001544