Order Entry
Czech Republic
ContactUsLinkComponent
4'-Methoxypropiophenone 99%
Irritant
4'-Methoxypropiophenone 99%
Catalog # APOSOR8854-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-Methoxypropiophenone 99%
Catalog # APOSOR8854-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :164.2 g/mol
Boiling Pt:273…275 °C
Melting Pt:27…29 °C
Density:1.07 g/cm³ (25 °C)
Flash Pt:61 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:121-97-1
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009310