

Tetrachloro-p-benzoquinone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:118-75-2
:
Formula:C₆Cl₄O₂
MDL Number:MFCD00001594
:245.88 g/mol
Melting Pt:287…290 °C
Density:1.97 g/cm³ (25 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:204-274-4
UN:3077
ADR:9+EHS,III
:GHS07
:GHS09
Frequently Bought Together