
5-Methoxyindole-3-acetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2436-17-1
:
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD02094163
:186.22 g/mol
Melting Pt:65…69 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together