
5-Morpholino-2-thiophenecarbaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24372-49-4
:
Formula:C₉H₁₁NO₂S
MDL Number:MFCD02710611
:197.26 g/mol
Melting Pt:126…128 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together