

2-Oxoindoline-5-sulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:199328-31-9
:
Formula:C₈H₆ClNO₃S
MDL Number:MFCD06655924
Melting Pt:160 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:231.66 g/mol
:GHS05
:GHS07
Frequently Bought Together