Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Dimethoxy-5-pyrimidineboronic acid
Irritant
2,4-Dimethoxy-5-pyrimidineboronic acid
Catalog # APOSOR8664-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dimethoxy-5-pyrimidineboronic acid
Catalog # APOSOR8664-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.9577 g/mol
Melting Pt:113…117 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:89641-18-9
Formula:C₆H₉BN₂O₄
MDL Number:MFCD01318985