Order Entry
Czech Republic
ContactUsLinkComponent
7-Methoxyisoquinoline 96%
Irritant
7-Methoxyisoquinoline 96%
Catalog # APOSOR8563-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Methoxyisoquinoline 96%
Catalog # APOSOR8563-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:48…50 °C
 :159.1900 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:39989-39-4
Formula:C₁₀H₉NO
MDL Number:MFCD09030747