1-[2-(Methylsulphonyl)phenyl]piperidine-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:942474-20-6
:
Formula:C₁₃H₁₇NO₄S
MDL Number:MFCD08692435
:283.35 g/mol
Melting Pt:195…198 °C
Storage Temperature:Ambient
Frequently Bought Together