
2-Chloro-4-(chloromethyl)thiazole
Catalog # APOSOR8403-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:168.05 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:5198-76-5
Formula:C₄H₃Cl₂NS
MDL Number:MFCD09265510